Thermo Scientific Chemicals

4-Methoxybenzyl carbazate, 97%, Thermo Scientific Chemicals

Catalog number: L00541.06
5 g, Each
Thermo Scientific Chemicals

4-Methoxybenzyl carbazate, 97%, Thermo Scientific Chemicals

Catalog number: L00541.06
5 g, Each
Quantity
Catalog number: L00541.06
also known as L00541-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18912-37-3
IUPAC Name
[(4-methoxyphenyl)methoxy]carbohydrazide
Molecular Formula
C9H12N2O3
InChI Key
JKBMMHKEAGEILO-UHFFFAOYSA-N
SMILES
COC1=CC=C(COC(=O)NN)C=C1
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to cream to pale brown

Description

It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text