Ethyl 3-bromopyruvate, tech. 75%
Ethyl 3-bromopyruvate, tech. 75%
Ethyl 3-bromopyruvate, tech. 75%
Thermo Scientific Chemicals

Ethyl 3-bromopyruvate, tech. 75%

CAS: 70-23-5 | C5H7BrO3 | 195.01 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
產品號碼 L00582.14
亦稱為 L00582-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS70-23-5
規格Specification Sheet規格表
Appearance (Color)Clear yellow to pink-red
Assay (GC)≥70.0%
CommentMay contain HBr gas
Identification (FTIR)Conforms
FormLiquid
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

Solubility
Difficult to mix.

Notes
Moisture Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Dale L. Boger.; Chen, J.H. A Modified Friedlander Condensation for the Synthesis of 3-Hydroxyquinoline-2-carboxylates. J. Org. Chem. 1995, 60, (22), 7369-7371.
  2. Brindaban C. Ranu.; Laksmikanta Adak.; Subhash Banerjee. Ionic liquid promoted interrupted Feist-Benary reaction with high diastereoselectivity. Tetrahedron Letters. 2008, 49, (31), 4613-4617.