5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%
5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%
5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%
5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%
Thermo Scientific Chemicals

5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%

CAS: 1136-45-4 | C11H9NO3 | 203.197 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L01142.14
also known as L01142-14
Price (TWD)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1136-45-4
IUPAC Name5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
Molecular FormulaC11H9NO3
InChI KeyPENHKTNQUJMHIR-UHFFFAOYSA-N
SMILESCC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)189.0-196.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to pale brown
Assay (GC)≥98.5%
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only