5-Aminoisoquinoline, 99%
5-Aminoisoquinoline, 99%
5-Aminoisoquinoline, 99%
Thermo Scientific Chemicals

5-Aminoisoquinoline, 99%

CAS: 1125-60-6 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L01223.03
also known as L01223-03
Price (EUR)
28,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
28,90
Each
Chemical Identifiers
CAS1125-60-6
IUPAC Nameisoquinolin-5-amine
Molecular FormulaC9H8N2
InChI KeyDTVYNUOOZIKEEX-UHFFFAOYSA-N
SMILESNC1=C2C=CN=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps or fused solid or granules
Appearance (Color)Pale cream to cream to yellow to orange to brown
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only