2,4,6-Tri-tert-butylaniline, 95%
2,4,6-Tri-tert-butylaniline, 95%
2,4,6-Tri-tert-butylaniline, 95%
2,4,6-Tri-tert-butylaniline, 95%
Thermo Scientific Chemicals

2,4,6-Tri-tert-butylaniline, 95%

CAS: 961-38-6 | C18H31N | 261.453 g/mol
製品番号(カタログ番号) L01362.06
または、製品番号L01362-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS961-38-6
IUPAC Name2,4,6-tri-tert-butylaniline
Molecular FormulaC18H31N
InChI KeyREJGDSCBQPJPQT-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(=C(N)C(=C1)C(C)(C)C)C(C)(C)C
さらに表示
Appearance (Color)White to cream
Assay (GC)≥94.0%
FormCrystals or powder or crystalline powder or fused solid
2,4,6-Tri-tert-butylnitrobenzene (bulky amine) is used in the synthesis of monomeric iminophosphorane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tri-tert-butylnitrobenzene (bulky amine) is used in the synthesis of monomeric iminophosphorane

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Ryusei Konaka.; Shigeru Terabe. Spin trapping of short-lived free radicals by use of 2,4,6-tri-tert-butylnitrosobenzene. J. Am. Chem. Soc. 1971 93, (17), 4306-4307.
  2. Guofu Zi.; Laura L. Blosch.; Li Jia.; Richard A. Andersen. Preparation and Reactions of Base-Free Bis(1,2,4-tri-tert-butylcyclopentadienyl)uranium Methylimide, Cp‘2UNMe, and Related Compounds. Organometallics. 2005, 24, (19), 4602-4612.