1,4-Benzodioxan-6-amine, 99%
1,4-Benzodioxan-6-amine, 99%
1,4-Benzodioxan-6-amine, 99%
Thermo Scientific Chemicals

1,4-Benzodioxan-6-amine, 99%

CAS: 22013-33-8 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L01421.06
also known as L01421-06
Price (EUR)
30,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
30,00
Each
Chemical Identifiers
CAS22013-33-8
IUPAC Name2,3-dihydro-1,4-benzodioxin-6-amine
Molecular FormulaC8H9NO2
InChI KeyBZKOZYWGZKRTIB-UHFFFAOYSA-N
SMILESNC1=CC=C2OCCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Pale yellow to yellow to dark red to dark brown to brown to black
FormCrystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Refractive Index1.5985-1.6015 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only