3,4,7,8-Tetramethyl-1,10-phenanthroline acts as a metal-chelating agent for transition metal-catalyzed processes. It is also used in the preparation of heteroleptic cationic iridium(III) complex viz. 3,4,7,8-tetramethyl-1,10-phenanthroline-bis[2-(2',4'-difluorophenyl)pyridine]iridium(III) hexafluorophosphate. Further, it is employed as a ligand and forms dinuclear copper(II) hypocrellin B complexes. In addition to this, it plays an important role in the formation of tetraaqua(3,4,7,8-tetramethyl-1,10-phenanthroline-kappa2N,N')zinc(II) thiosulfate complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El 3,4,7,8-tetrametil-1,10-fenantrolina actúa como agente quelante para procesos de transición catalizada por metales. También se utiliza en la preparación del complejo heteroléptico catiónico iridio(III), es decir, 3,4,7,8-tetrametilo-1,10-fenantrolina-bis[2-(2′,4′-difluorofenil)piridina]iridio(III) hexafluorofosfato. Asimismo, se emplea como ligando y forma complejos de hipocretina B de cobre dinuclear(II). Además, desempeña un papel importante en la formación del complejo tetraaqua(3,4,7,8-tetrametil-1,10-fenantrolina-kappa2N,N′)cinc(II) tiosulfato.
Solubilidad
Soluble en 1,4-dioxano, acetona, diclorometano, N,N-dimetilformamida, N,N-dimetilacetamida, metanol y tolueno caliente.
Notas
Incompatible con los agentes oxidantes.
RUO – Research Use Only
General References:
- Hanifehpour, Y.; Mirtamizdoust, B.; Morsali, A.; Joo, S. W. Sonochemical syntheses of binuclear lead(II)-azido supramolecule with ligand 3,4,7,8-tetramethyl-1,10-phenanthroline as precursor for preparation of lead(II) oxide nanoparticles. Ultrason. Sonochem. 2015, 23, 275-281.
- Tong, Y. Z.; Gao, C.; Wang, Q. L.; Wang, B. W.; Gao, S.; Cheng, P.; Liao, D. Z. Two mononuclear single molecule magnets derived from dysprosium(III) and tmphen (tmphen = 3,4,7,8-tetramethyl-1,10-phenanthroline). Dalton Trans. 2015, 44 (19), 9020-9026.