Norcamphor, 98+%
Norcamphor, 98+%
Norcamphor, 98+%
Thermo Scientific Chemicals

Norcamphor, 98+%

Used as a building block in organic synthesis and also acts as a precursor to norborneols. | CAS: 497-38-1 | C7H10O | 110.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01491.14
also known as L01491-14
Price (USD)
106.65
Special offer
Online exclusive
Ends: 30-Jun-2026
125.00
Save 18.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 30-Jun-2026
125.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS497-38-1
IUPAC Namebicyclo[2.2.1]heptan-2-one
Molecular FormulaC7H10O
InChI KeyKPMKEVXVVHNIEY-UHFFFAOYNA-N
SMILESO=C1CC2CCC1C2
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)86-99?C
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)Colorless to white to cream
FormWaxy crystalline solid or compacted crystalline powder
View more
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
RUO – Research Use Only

General References:

  1. Jiang, X.; Pan, Z.; Douglas, C. J. Cyclization of an alkene-bearing cyclopentanone: The role of rhodium and Brønsted acid. Tetrahedron Lett. 2015, 56 (39), 5324-5327.
  2. Xu, W.; Langer, R. Probing the effect of heterocycle-bonding in PNX-type ruthenium pre-catalysts for reactions involving H2. Dalton Trans 2015, 44 (38), 16785-16790.