Thermo Scientific Chemicals

Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol, Thermo Scientific Chemicals

Catalog number: L01519.03
1 g, Each
Thermo Scientific Chemicals

Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol, Thermo Scientific Chemicals

Catalog number: L01519.03
1 g, Each
Quantity
Catalog number: L01519.03
also known as L01519-03
Price (USD)
Price: 32.10
Online price: 27.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3680-02-2
IUPAC Name
methanesulfonylethene
Molecular Formula
C3H6O2S
InChI Key
WUIJTQZXUURFQU-UHFFFAOYSA-N
SMILES
CS(=O)(=O)C=C
Solution Test
10% v/v in methanol: Clear
Appearance (Color)
Clear, colorless to pale yellow
Assay (GC)
>94.0%
Refractive Index
1.4610-1.4650 @ 20?C
Solution Test
10% v/v in diethyl ether: Clear

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text