Ethyl 3-dimethylaminopropionate, 97%
Ethyl 3-dimethylaminopropionate, 97%
Ethyl 3-dimethylaminopropionate, 97%
Thermo Scientific Chemicals

Ethyl 3-dimethylaminopropionate, 97%

CAS: 20120-21-2 | C7H15NO2 | 145.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01542.06
also known as L01542-06
Price (EUR)
26,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
26,00
Each
Chemical Identifiers
CAS20120-21-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Refractive Index1.4165-1.4215 @ 20?C
Assay (GC)≥96.0%
Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Alkyl 3-dimethylaminopropionates have been used as the synthetic equivalents of acrylate anions, by ɑ-lithiation, reaction with the appropriate electrophile and elimination: J. Org. Chem., 46, 4536 (1981):
  2. Vanderveen, J. R.; Durelle, J.; Jessop, P. G. Design and evaluation of switchable-hydrophilicity solvents. Green Chem. 2014, 16, 1187-1197.
  3. Zhi, Y.; Gao, L. X.; Jin, Y.; Tang, C. L.; Li, J. Y.; Li, J.; Long, Y. Q. 4-Quinolone-3-carboxylic acids as cell-permeable inhibitors of protein tyrosine phosphatase 1B. Bioorg. Med. Chem. 2014, 22 (14), 3670-3683.