Acetal dimetílico de N,N-dimetilacetamida, téc. 90 %, estabilizado con 5-10 % metanol, Thermo Scientific Chemicals
Acetal dimetílico de N,N-dimetilacetamida, téc. 90 %, estabilizado con 5-10 % metanol, Thermo Scientific Chemicals
Acetal dimetílico de N,N-dimetilacetamida, téc. 90 %, estabilizado con 5-10 % metanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetal dimetílico de N,N-dimetilacetamida, téc. 90 %, estabilizado con 5-10 % metanol, Thermo Scientific Chemicals

CAS: 18871-66-4 | C6H15NO2 | 133.191 g/mol
Have Questions?
Número de catálogoCantidad
L01642.14
también denominado L01642-14
25 g
Número de catálogo L01642.14
también denominado L01642-14
Precio (USD)
-
Cantidad:
25 g
Pedido a granel o personalizado
Identificadores químicos
CAS18871-66-4
IUPAC Name(1,1-dimethoxyethyl)dimethylazanium
Molecular FormulaC6H16NO2
InChI KeyFBZVZUSVGKOWHG-UHFFFAOYSA-O
SMILESCOC(C)(OC)[NH+](C)C
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
StabilizerSodium carbonate (Na2CO3): 0.1-0.5% by weight (non-U.S. specification)
Appearance (Color)Clear colorless to yellow
Assay (Non-aqueous acid-base Titration)≥88.0% (non-U.S. specification)
Assay from Suppliers CofA≥88.0% (Proton-NMR, U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Ver más
N,N-Dimethylacetamide dimethyl acetal is used as a reagent for the synthesis of amides, diacylamines, and heterocycles, one carbon inserting synthon in preparation of pyrimido[1,2-a][1,3,5]triazin-6-ones. It is used as intermediate in building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El acetal dimetílico de N,N-dimetilacetamida se utiliza como reactivo para la síntesis de amidas, diacilaminas y heterociclos, un sintón con inserto de carbono en una preparación de pirimido[1,2-a][1,3,5]triazin-6-onas. También se usa como intermedio en compuestos básicos.

Solubilidad
Hidroliza en agua.

Nota
Sensible a la humedad. Almacenar a temperaturas entre los 2 y los 8 °C. Incompatible con agentes oxidantes fuertes y ácidos.
RUO – Research Use Only

General References:

  1. Richard K. Hill; Raghovan. Soman; Seiji. Sawada. Asymmetric induction in the thermal reactions of allylic alcohols with N,N-dimethylacetamide dimethylacetal and triethyl orthoacetate.J. Org. Chem.1972, 37 (23), 3737-3740.
  2. Robert G. Salomon, Swadesh R. Raychaudhuri. Convenient preparation of N,N-dimethylacetamide dimethyl acetal.J. Org. Chem.1984, 49 (19), 3659-3660.
  3. Reacts with 1,2-diols, e.g. sugars, to form 1-(dimethylamino)ethylidene cyclic acetals which can be cleaved with aqueous AcOH, giving the monoacetate of the less hindered OH, thus providing a method for selective monoprotection of the diol: Can. J. Chem., 50, 233 (1972).
  4. Reacts with active methylene compounds to give the (dimethylamino)ethylidene derivatives: Liebigs Ann. Chem., 641, 1 (1961), compare N,N-Dimethyl formamide dimethyl acetal, A15350.
  5. For use in the N-acetylation of amides to give diacylamines (imides) in high yield, see: Synthesis, 119 (1980).