5,6-Dihydrouracil, 97%
5,6-Dihydrouracil, 97%
5,6-Dihydrouracil, 97%
Thermo Scientific Chemicals

5,6-Dihydrouracil, 97%

CAS: 504-07-4 | C4H6N2O2 | 114.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L01918.03
also known as L01918-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS504-07-4
IUPAC Name1,3-diazinane-2,4-dione
Molecular FormulaC4H6N2O2
InChI KeyOIVLITBTBDPEFK-UHFFFAOYSA-N
SMILESO=C1CCNC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

Solubility
Soluble in Sodium Hydroxide.

Notes
Incompatible with oxidizing agents.Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. David Kupfer; Ruth Partridge. 6β-Hydroxylation of triamcinolone acetonide by a hepatic enzyme system: The effect of phenobarbital and 1-benzyl-2-thio-5,6-dihydrouracil.Archives of Biochemistry and Biophysics.1970, 140 23-28.
  2. André B.P. van Kuilenburga; Henk van Lenthe; Arno van Cruchten and Willem Kulik. Quantification of 5,6-Dihydrouracil by HPLC-Electrospray Tandem Mass Spectrometry.Clinical Chemistry.2004, 50 236-238.