Phenanthrene, 98%
Phenanthrene, 98%
Phenanthrene, 98%
Thermo Scientific Chemicals

Phenanthrene, 98%

CAS: 85-01-8 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L01921.18
also known as L01921-18
Price (EUR)
59,60
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
59,60
Each
Add to cart
Chemical Identifiers
CAS85-01-8
IUPAC Namephenanthrene
Molecular FormulaC14H10
InChI KeyYNPNZTXNASCQKK-UHFFFAOYSA-N
SMILESC1=CC=C2C(C=CC3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to pale brown or grey
FormCrystals or powder or crystalline powder or flakes
Identification (FTIR)Conforms
Melting Point (clear melt)97-101°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

Solubility
Soluble in Ether, Benzene, Chloroform, Toluene. Insoluble in water.

Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. T. Azumi and S. P. McGlynn. Polarization of the Luminescence of Phenanthrene.J. Chem. Phys.1962, 37 2413.
  2. Yanzheng Gao; Lizhong Zhu. Plant uptake, accumulation and translocation of phenanthrene and pyrene in soils.Chemosphere.2004, 55 1169-1178.