Isopentylamine, 99%
Isopentylamine, 99%
Isopentylamine, 99%
Thermo Scientific Chemicals

Isopentylamine, 99%

CAS: 107-85-7 | C5H13N | 87.166 g/mol
Have Questions?
Catalog NumberQuantity
L02260.14
also known as L02260-14
25 g
Catalog number L02260.14
also known as L02260-14
Price (EUR)
46,80
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
46,80
Each
Add to cart
Chemical Identifiers
CAS107-85-7
IUPAC Name3-methylbutan-1-amine
Molecular FormulaC5H13N
InChI KeyBMFVGAAISNGQNM-UHFFFAOYSA-N
SMILESCC(C)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4065-1.4100 @ 20°C (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only