Thermo Scientific Chemicals

Benzo[b]furan-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: L02456.04
2 g, Each
Thermo Scientific Chemicals

Benzo[b]furan-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: L02456.04
2 g, Each
Quantity
Catalog number: L02456.04
also known as L02456-04
Price (USD)
Price: 116.00
Online price: 99.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4265-16-1
IUPAC Name
1-benzofuran-2-carbaldehyde
Molecular Formula
C9H6O2
InChI Key
ADDZHRRCUWNSCS-UHFFFAOYSA-N
SMILES
O=CC1=CC2=CC=CC=C2O1
Assay (GC)
>98.5%
Free acid (titration)
≤1.0%
Refractive Index
1.6300-1.6345 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text