trans-4-(Trifluoromethyl)cinnamic acid, 98%
trans-4-(Trifluoromethyl)cinnamic acid, 98%
trans-4-(Trifluoromethyl)cinnamic acid, 98%
trans-4-(Trifluoromethyl)cinnamic acid, 98%
Thermo Scientific Chemicals

trans-4-(Trifluoromethyl)cinnamic acid, 98%

CAS: 16642-92-5 | C10H6F3O2 | 215.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L02771.06
also known as L02771-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS16642-92-5
IUPAC Name(2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate
Molecular FormulaC10H6F3O2
InChI KeyANRMAUMHJREENI-ZZXKWVIFSA-M
SMILES[O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only