3,4,5-Trimethylphenol, 97%
3,4,5-Trimethylphenol, 97%
3,4,5-Trimethylphenol, 97%
Thermo Scientific Chemicals

3,4,5-Trimethylphenol, 97%

CAS: 527-54-8 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L02911.06
also known as L02911-06
Price (EUR)
167,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
167,00
Each
Chemical Identifiers
CAS527-54-8
IUPAC Name3,4,5-trimethylphenol
Molecular FormulaC9H12O
InChI KeyFDQQNNZKEJIHMS-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to cream to yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only