3-Phenylpropylamine, 98+%
3-Phenylpropylamine, 98+%
3-Phenylpropylamine, 98+%
Thermo Scientific Chemicals

3-Phenylpropylamine, 98+%

CAS: 2038-57-5 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L03023.18
also known as L03023-18
Price (EUR)
104,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS2038-57-5
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Refractive Index1.5240-1.5280 @ 20?C
Assay (GC)≥98.0%
FormLiquid
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.

Solubility
Soluble in Chloroform, Ethyl Acetate. Not miscible in water.

Notes
Store away from air and under dry inert gas. Air sensitive. Incompatible with acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
RUO – Research Use Only

General References:

  1. Arslan Ünala; Mustafa Senyela; Sükrü Sentürk. Conformational analysis and vibrational spectroscopic investigation of 3-phenylpropylamine.Vibrational Spectroscopy.2009, 50 277-284.
  2. Mustafa Senyela; Arslan Unal; Ozgür Alver. Molecular structure, NMR analyses, density functional theory and ab initio Hartree-Fock calculations of 3-phenylpropylamine.Comptes Rendus Chimie.2009, 12 808-815.