Thermo Scientific Chemicals

3-Methyl-4-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog number: L03163.14
25 g, Each
Thermo Scientific Chemicals

3-Methyl-4-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog number: L03163.14
25 g, Each
Quantity
Catalog number: L03163.14
also known as L03163-14
Price (USD)
Price: 43.90
Online price: 34.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2581-34-2
Assay from Supplier's CofA
≥97.5% (HPLC, U.S. sourced material)
Melting Point (clear melt)
125.0-131.0°C (non-U.S. sourced material)
Form
Crystals or powder or crystalline powder
Appearance (Color)
Cream to brown to yellow
Assay (GC)
≥97.5% (non-U.S. sourced material)

Description

3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

Solubility
Soluble in water(1.34g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text