Thermo Scientific Chemicals

Benzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: L03292.0E
2500 g, Each
Thermo Scientific Chemicals

Benzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: L03292.0E
2500 g, Each
Quantity
Catalog number: L03292.0E
also known as L03292-0E
Price (USD)
Price: 138.00
Online price: 117.65
Your price:
Quantity
-

Chemical Identifiers

CAS
100-51-6
IUPAC Name
phenylmethanol
Molecular Formula
C7H8O
InChI Key
WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5380-1.5415 @ 20°C

Description

Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

Solubility
Misicible with water, benzene, methanol, chloroform, ethanol, acetone and ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text