Allyl phenyl ether, 99%
Allyl phenyl ether, 99%
Allyl phenyl ether, 99%
Thermo Scientific Chemicals

Allyl phenyl ether, 99%

CAS: 1746-13-0 | C9H10O | 134.178 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
產品號碼 L03359.14
亦稱為 L03359-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS1746-13-0
IUPAC Name(prop-2-en-1-yloxy)benzene
Molecular FormulaC9H10O
InChI KeyPOSICDHOUBKJKP-UHFFFAOYSA-N
SMILESC=CCOC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear, colourless
Formliquid
Assay (GC)>98.5%
Refractive Index1.5200-1.5230
Allyl phenyl ether is an pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl phenyl ether is an pharmaceutical and OLED intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents, acids, bases. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Peter Golborn and Feodor Scheinmann. Isomerisation of allyl phenyl ethers and allylphenols with transition metal catalysts. J. Chem. Soc., Perkin Trans. 1. 1973, 2870-2875.
  2. Eric E. Kingston; John H. Beynon; Joachim G. Liehr; Philippe Meyrant; Robert Flammang and André Maquestiau. The Claisen rearrangement of protonated allyl phenyl ether. Organic Mass Spectrometry. 1989, 20, (5), 351-359.