Dimethylcarbamyl chloride, 96%
Dimethylcarbamyl chloride, 96%
Dimethylcarbamyl chloride, 96%
Thermo Scientific Chemicals

Dimethylcarbamyl chloride, 96%

CAS: 79-44-7 | C3H6ClNO | 107.537 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L03415.14
also known as L03415-14
Price (EUR)
22,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
22,00
Each
Chemical Identifiers
CAS79-44-7
IUPAC NameN,N-dimethylcarbamoyl chloride
Molecular FormulaC3H6ClNO
InChI KeyYIIMEMSDCNDGTB-UHFFFAOYSA-N
SMILESCN(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4500-1.4560 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
Identification (FTIR)Conforms
FormLiquid
Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

Solubility
Reacts with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
RUO – Research Use Only
Gyorgy Schultz; Istvan Hargittai. Molecular Structure of Dimethylcarbamyl Chloride from Gas-Phase Electron Diffraction.J. Phys. Chem.1995, 99 (29), 11412-11416.