Dimethylsilyldiethylamine, 95%
Dimethylsilyldiethylamine, 95%
Dimethylsilyldiethylamine, 95%
Thermo Scientific Chemicals

Dimethylsilyldiethylamine, 95%

CAS: 13686-66-3 | C6H16NSi | 130.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03503.06
also known as L03503-06
Price (USD)
62.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.80
Each
Chemical Identifiers
CAS13686-66-3
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to standard
Refractive Index1.4080
Appearance (Color)Colorless
FormLiquid
Assay (GC)≥94%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only