n-Hexyltrimethoxysilane, 97%
n-Hexyltrimethoxysilane, 97%
n-Hexyltrimethoxysilane, 97%
Thermo Scientific Chemicals

n-Hexyltrimethoxysilane, 97%

CAS: 3069-19-0 | C9H22O3Si | 206.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03576.06
also known as L03576-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3069-19-0
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Refractive Index1.4055-1.4105 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only