Iodoacetonitrile, 97%
Iodoacetonitrile, 97%
Iodoacetonitrile, 97%
Thermo Scientific Chemicals

Iodoacetonitrile, 97%

CAS: 624-75-9 | C2H2IN | 166.949 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L03634.09
also known as L03634-09
Price (TWD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS624-75-9
IUPAC Name2-iodoacetonitrile
Molecular FormulaC2H2IN
InChI KeyVODKOOOHHCAWFR-UHFFFAOYSA-N
SMILESICC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to orange to red or brown
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5740-1.5790 @ 20?C
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.

Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.
RUO – Research Use Only

General References:

  1. Paul G. Wenthold. Determination of the proton affinities of bromo- and iodoacetonitrile using the kinetic method with full entropy analysis.Journal of the American Society for Mass Spectrometry.2000, 11 601-605.
  2. W. C. Kupferschmidt; R. B. Jordan. Reactions of chromium(II) with chloro- and iodoacetonitrile and their pentaamminecobalt(III) complexes.J. Am. Chem. Soc..1984, 106 (4), 991-995.