Methyl 2-hydroxy-4-methoxybenzoate, 98%
Methyl 2-hydroxy-4-methoxybenzoate, 98%
Methyl 2-hydroxy-4-methoxybenzoate, 98%
Methyl 2-hydroxy-4-methoxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-hydroxy-4-methoxybenzoate, 98%

CAS: 5446-02-6 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L03855.18
also known as L03855-18
Price (TWD)
-
Request A Quote
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS5446-02-6
IUPAC Namemethyl 2-hydroxy-4-methoxybenzoate
Molecular FormulaC9H10O4
InChI KeyZICRWXFGZCVTBZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(OC)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)47.0-53.0?C
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Wenjing Wang; Jijun Xue; Tian Tian; Yingdong Jiao; Ying Li. The first asymmetric total synthesis of (+)-coriandrone A and B.Organic & Biomolecular Chemistry.2013, 11 (39), 6686-6690.