Ethyl phenylcyanoacetate, 98%
Ethyl phenylcyanoacetate, 98%
Ethyl phenylcyanoacetate, 98%
Thermo Scientific Chemicals

Ethyl phenylcyanoacetate, 98%

CAS: 4553-07-5 | C11H11NO2 | 189.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 L03929.06
亦稱為 L03929-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS4553-07-5
IUPAC Nameethyl 2-cyano-2-phenylacetate
Molecular FormulaC11H11NO2
InChI KeySXIRJEDGTAKGKU-UHFFFAOYNA-N
SMILESCCOC(=O)C(C#N)C1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.5025-1.5075 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only