Thermo Scientific Chemicals

N,N,N',N'-Tetramethylmethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: L04037.22
100 g, Each
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylmethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: L04037.22
100 g, Each
Quantity
Catalog number: L04037.22
also known as L04037-22
Price (USD)
Price: 164.00
Online price: 141.65
Your price:
Quantity
-

Chemical Identifiers

CAS
51-80-9
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.3995-1.4025 @ 20?C

Description

N,N,N',N'-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text