Thermo Scientific Chemicals

4-Chloro-1-butanol, tech. 85%, Thermo Scientific Chemicals

Catalog number: L04042.30
250 g, Each
Thermo Scientific Chemicals

4-Chloro-1-butanol, tech. 85%, Thermo Scientific Chemicals

Catalog number: L04042.30
250 g, Each
Quantity
Catalog number: L04042.30
also known as L04042-30
Price (USD)
Price: 244.00
Online price: 207.65
Your price:
Quantity
-

Chemical Identifiers

CAS
928-51-8
Appearance (Color)
Clear to slightly hazy colorless to yellow or pale pink
Assay (GC)
≥80.0%
Refractive Index
1.4510-1.4630 @ 20°C
Free acid (titration)
≤6.0% as HCl
Form
Liquid

Description

4-Chloro-1-butanol is employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1-butanol is employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

Solubility
Soluble in methanol, chloroform, and dimethyl sufoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text