Castor oil
Castor oil
Castor oil
Thermo Scientific Chemicals

Castor oil

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
2500 mL
Catalog number L04224.AU
also known as L04224-AU
Price (EUR)
61,70
Each
Quantity:
1000 mL
Request bulk or custom format
Price (EUR)
61,70
Each
Chemical Identifiers
CAS8001-79-4
IUPAC Name1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
Molecular FormulaC57H104O9
InChI KeyZEMPKEQAKRGZGQ-UHFFFAOYNA-N
SMILESCCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4760-1.4810 @ 20?C
FormViscous liquid
Appearance (Color)Clear colorless to yellow
Optical Rotation+4.5 ? 1?(c=5 in ethanol)
Castor oil is used in food additives, flavorings, candy and as a mold inhibitor. Its derivatives are used in the production of soaps, lubricants, hydraulic and brake fluids, paints, coatings, lithium grease and cold resistant plastics. It finds application in waxes, polishes, nylon, pharmaceuticals and perfumes. It is involved in the preparation of linoleic acids by dehydration, 2-octanol, sebacic acid and undecylenic acid, which is a precursor for polymer nylon 11 and heptanal. It is involved in the preparation of 12-hydroxystearic acid by hydrogenation and saponification. It is useful as a lubricant in jet, diesel, and racing engine. Further, it is used as a laxative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Castor oil is used in food additives, flavorings, candy and as a mold inhibitor. Its derivatives are used in the production of soaps, lubricants, hydraulic and brake fluids, paints, coatings, lithium grease and cold resistant plastics. It finds application in waxes, polishes, nylon, pharmaceuticals and perfumes. It is involved in the preparation of linoleic acids by dehydration, 2-octanol, sebacic acid and undecylenic acid, which is a precursor for polymer nylon 11 and heptanal. It is involved in the preparation of 12-hydroxystearic acid by hydrogenation and saponification. It is useful as a lubricant in jet, diesel, and racing engine. Further, it is used as a laxative.

Solubility
Miscible with methanol, diethyl ether, chloroform, glacial acetic acid, benzene and carbon disulfide. Immiscible with cold water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, acids and bases.
RUO – Research Use Only

General References:

  1. Gurunathan, T.; Mohanty, S.; Nayak, S. K. Isocyanate terminated castor oil-based polyurethane prepolymer: Synthesis and characterization. Prog. Org. Coat. 2015, 80, 39-48.
  2. Saez-Bastante, J.; Pinzi, S.; Jimenez-Romero, F. J.; de Castro, M. D. L.; Priego-Capote, F.; Dorado, M. P. Synthesis of biodiesel from castor oil: Silent versus sonicated methylation and energy studies. Energy Convers. Manage. 2015, 96, 561-567.