Ethyl hydrogen suberate, 98%
Ethyl hydrogen suberate, 98%
Ethyl hydrogen suberate, 98%
Ethyl hydrogen suberate, 98%
Thermo Scientific Chemicals

Ethyl hydrogen suberate, 98%

CAS: 14113-01-0 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04272.14
also known as L04272-14
Price (TWD)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS14113-01-0
IUPAC Name8-ethoxy-8-oxooctanoic acid
Molecular FormulaC10H18O4
InChI KeyKCFVQEAPUOVXTH-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Refractive Index1.4415-1.4455 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only