Description
1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .
Solubility
Soluble in water, soluble in chloroform
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only