4-Bromoheptane, 97%
4-Bromoheptane, 97%
4-Bromoheptane, 97%
Thermo Scientific Chemicals

4-Bromoheptane, 97%

CAS: 998-93-6 | C7H15Br | 179.10 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 L04807.14
亦稱為 L04807-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS998-93-6
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4440-1.4505 @ 20?C
4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

Solubility
Miscible with carbon tetrachloride, chloroform and benzene. Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Molander, G. A.; Argintaru, O. A. Stereospecific Ni-Catalyzed Cross-Coupling of Potassium Alkenyltrifluoroborates with Alkyl Halides. Org. Lett. 2014, 16 (7), 1904-1907.
  2. Kim, S. W.; Hooker, J. M.; Otto, N.; Win, K.; Muench, L.; Shea, C.; Carter, P.; King, P.; Reid, A. E.; Volkow, N. D.; Fowler, J. S. Whole-body pharmacokinetics of HDAC inhibitor drugs, butyric acid, valproic acid and 4-phenylbutyric acid measured with carbon-11 labeled analogs by PET. Nucl. Med. Biol. 2013, 40 (7), 912-918.