Thermo Scientific Chemicals

Dibenzyl phthalate, 97%, Thermo Scientific Chemicals

5g, Each
Thermo Scientific Chemicals

Dibenzyl phthalate, 97%, Thermo Scientific Chemicals

5g, Each
Quantity
Catalog number: L04985.06
also known as L04985-06
Price (USD)
Price: 115.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
523-31-9
IUPAC Name
1,2-dibenzyl benzene-1,2-dicarboxylate
Molecular Formula
C22H18O4
InChI Key
UCVPKAZCQPRWAY-UHFFFAOYSA-N
SMILES
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Form
Crystals or powder or crystalline powder or lumps or fused solid
Melting Point (clear melt)
38.0-48.0?C
Appearance (Color)
White to pale cream to pale brown
Assay (GC)
≥96.0%

Description

Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.

Solubility
Insoluble in water.

Notes
Incompatible materials are reducing agents. Strong acids. Strong bases and Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only