Thermo Scientific Chemicals

1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: L05336.14
25 g, Each
Thermo Scientific Chemicals

1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: L05336.14
25 g, Each
Quantity
Catalog number: L05336.14
also known as L05336-14
Price (USD)
Price: 286.00
Online price: 246.65
Your price:
Quantity
-

Chemical Identifiers

CAS
58880-37-8
IUPAC Name
1-(4-chlorophenyl)cyclohexane-1-carboxylate
Molecular Formula
C13H14ClO2
InChI Key
UPNXUJXIIZGXLQ-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Appearance (Color)
White to grey or purple
Form
Powder or granules
Identification (FTIR)
Conforms
Melting Point (clear melt)
146.5-158.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text