Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads
Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads
Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads
Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads
Thermo Scientific Chemicals

Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number L05722.0B
also known as L05722-0B
Price (EUR)
126,00
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
126,00
Each
Add to cart
Chemical Identifiers
CAS69912-79-4
IUPAC NameMolecular sieves 5A
MDL NumberMFCD00147627
SpecificationsSpecification SheetSpecification Sheet
CommentVolatiles ≤4.0%
Appearance (Color)Cream to pale brown
FormBeads
CommentWater adsorption capacity ≥14.5%
Molecular sieves, 13X, 3-5mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only