2-Amino-5-ciclopropil-1,3,4-tiadiazol, 98 %, Thermo Scientific Chemicals
2-Amino-5-ciclopropil-1,3,4-tiadiazol, 98 %, Thermo Scientific Chemicals
2-Amino-5-ciclopropil-1,3,4-tiadiazol, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-ciclopropil-1,3,4-tiadiazol, 98 %, Thermo Scientific Chemicals

CAS: 57235-50-4 | C5H7N3S | 141.19 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo L05778.03
también denominado L05778-03
Precio (USD)
-
Cantidad:
1 g
Pedido a granel o personalizado
Identificadores químicos
CAS57235-50-4
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Off-white to pale cream
Mass SpecConforms
FormPowder or crystals or crystalline powder
Purity≥97.5%
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El 2-amino-5-ciclopropil-1,3,4-tiadiazol se utiliza como agente antibacteriano.

Solubilidad
Soluble en metanol. Insoluble en agua.

Notas
Incompatible con agentes oxidantes. Almacenar en un lugar fresco y seco dentro de recipientes bien sellados. Almacenar a -20 °C.
RUO – Research Use Only

General References:

  1. Harun Patel; Lishu Mishra; Malleshappa Noolvi; Rajshekhar Karpoormath and Swaranjit Singh Cameotra. Synthesis, In Vitro Evaluation, and Molecular Docking Studies of Azetidinones and Thiazolidinones of 2-Amino-5-cyclopropyl-1,3,4-thiadiazole as Antibacterial Agents. Archiv der Pharmazie. 2014, 347, 668-684.
  2. Rohit Bhatia; Amit Sharma; Ankaj Kaundal. A Review on 1, 3, 4-Thiadiazole Derivatives. Indian Journal of Pharmaceutical Science & Research. 2014, 4, 165-172.