1,2-Ethylenediphosphonic acid, 97%
1,2-Ethylenediphosphonic acid, 97%
1,2-Ethylenediphosphonic acid, 97%
Thermo Scientific Chemicals

1,2-Ethylenediphosphonic acid, 97%

CAS: 6145-31-9 | C2H8O6P2 | 190.028 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05828.06
also known as L05828-06
Price (EUR)
94,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
94,20
Each
Chemical Identifiers
CAS6145-31-9
IUPAC Name(2-phosphonoethyl)phosphonic acid
Molecular FormulaC2H8O6P2
InChI KeyXYJLPCAKKYOLGU-UHFFFAOYSA-N
SMILESOP(O)(=O)CCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to cream
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only