O-(4-Nitrobenzyl)hydroxylamine hydrochloride, 98%
O-(4-Nitrobenzyl)hydroxylamine hydrochloride, 98%
O-(4-Nitrobenzyl)hydroxylamine hydrochloride, 98%
Thermo Scientific Chemicals

O-(4-Nitrobenzyl)hydroxylamine hydrochloride, 98%

CAS: 2086-26-2 | C7H9ClN2O3 | 204.61 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 L05861.03
亦稱為 L05861-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS2086-26-2
IUPAC Namehydrogen O-[(4-nitrophenyl)methyl]hydroxylamine chloride
Molecular FormulaC7H9ClN2O3
InChI KeyLKCAFSOYOMFQSL-UHFFFAOYSA-N
SMILES[H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only