1-Methylpyrrole-2-carboxylic acid, 98%
1-Methylpyrrole-2-carboxylic acid, 98%
1-Methylpyrrole-2-carboxylic acid, 98%
Thermo Scientific Chemicals

1-Methylpyrrole-2-carboxylic acid, 98%

CAS: 6973-60-0 | C6H7NO2 | 125.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05906.06
also known as L05906-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6973-60-0
IUPAC Name1-methyl-1H-pyrrole-2-carboxylic acid
Molecular FormulaC6H7NO2
InChI KeyILAOVOOZLVGAJF-UHFFFAOYSA-N
SMILESCN1C=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown to gray
FormCrystals or powder or crystalline powder or lumps
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only
Ana Filipa L O M Santos; Manuel A V Ribeiro da Silva. Experimental and computational study on the molecular energetics of 2-pyrrolecarboxylic acid and 1-methyl-2-pyrrolecarboxylic acid. Journal of Physical Chemistry A: Molecules, Spectroscopy, Kinetics, Environment and General Theory.2009, 113, (35), 9741-9750.