4-Tritylphenol, 98%
4-Tritylphenol, 98%
4-Tritylphenol, 98%
Thermo Scientific Chemicals

4-Tritylphenol, 98%

CAS: 978-86-9 | C25H20O | 336.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06073.06
also known as L06073-06
Price (EUR)
58,65
Online Exclusive
65,30
Save 6,65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
58,65
Online Exclusive
65,30
Save 6,65 (10%)
Each
Chemical Identifiers
CAS978-86-9
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Melting Point (clear melt)276-286°C
Appearance (Color)White to cream
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only