3',4',5'-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La 3′,4′,5′-trimetoxiacetofenona se utiliza en los campos agroquímicos, farmacéuticos y de colorantes.
Solubilidad
Ligeramente soluble en agua.
Notas
Los materiales incompatibles son los agentes oxidantes fuertes y las bases fuertes. Almacenar alejado de agentes oxidantes fuertes y bases fuertes. Evitar el calor excesivo y la exposición a la luz.
RUO – Research Use Only
General References:
- Y Engelborghs; C Dumortier; A D'Hoore; A Vandecandelaere; T J Fitzgerald. Evidence for an alternative pathway for colchicine binding to tubulin, based on the binding kinetics of the constituent rings..Journal of Biological Chemistry.1993,268 (1), 107-112.