DL-Valine methyl ester hydrochloride, 99%
DL-Valine methyl ester hydrochloride, 99%
DL-Valine methyl ester hydrochloride, 99%
DL-Valine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals

DL-Valine methyl ester hydrochloride, 99%

CAS: 5619-05-6 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06526.14
also known as L06526-14
Price (EUR)
147,05
Online Exclusive
173,00
Save 25,95 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
147,05
Online Exclusive
173,00
Save 25,95 (15%)
Each
Add to cart
Chemical Identifiers
CAS5619-05-6
IUPAC Namemethyl 2-amino-3-methylbutanoate hydrochloride
Molecular FormulaC6H14ClNO2
InChI KeyKUGLDBMQKZTXPW-UHFFFAOYSA-N
SMILESCl.COC(=O)C(N)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only