Thermo Scientific Chemicals

trans-1,2-Cyclohexanediol, 98%, Thermo Scientific Chemicals

Catalog number: L07088.14
25 g, Each
Thermo Scientific Chemicals

trans-1,2-Cyclohexanediol, 98%, Thermo Scientific Chemicals

Catalog number: L07088.14
25 g, Each
Quantity
Catalog number: L07088.14
also known as L07088-14
Price (USD)
Price: 79.00
Online price: 67.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1460-57-7
IUPAC Name
(1R,2R)-cyclohexane-1,2-diol
Molecular Formula
C6H12O2
InChI Key
PFURGBBHAOXLIO-PHDIDXHHSA-N
SMILES
O[C@@H]1CCCC[C@H]1O
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
99.0-106.0?C
Appearance (Color)
White to cream
Assay (GC)
≥97.5%

Description

trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text