(S)-(+)-Carvone, 96%
(S)-(+)-Carvone, 96%
(S)-(+)-Carvone, 96%
Thermo Scientific Chemicals

(S)-(+)-Carvone, 96%

CAS: 2244-16-8 | C10H14O | 150.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07130.14
also known as L07130-14
Price (USD)
66.40
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
66.40
Each
Chemical Identifiers
CAS2244-16-8
IUPAC Name(5S)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Molecular FormulaC10H14O
InChI KeyULDHMXUKGWMISQ-VIFPVBQESA-N
SMILESCC(=C)[C@H]1CC=C(C)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Optical Rotation58.0 ± 2.0? (neat)
Refractive Index1.4955-1.5030 @ 20?C
Appearance (Color)Clear colorless to white to yellow
FormLiquid
View more
Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

Solubility
Miscible with diethyl ether, ethanol and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Senthil Kumaran, R.; Mehta, G. A versatile, RCM based approach to eudesmane and dihydroagarofuran sesquiterpenoids from (-)-carvone: a formal synthesis of (-)-isocelorbicol. Tetrahedron. 2015, 71 (19), 1718-1731.
  2. Huchelmann, A.; Gastaldo, C.; Veinante, M.; Zeng, Y.; Heintz, D.; Tritsch, D.; Schaller, H.; Rohmer, M.; Bach, T. J.; Hemmerlin, A. S-Carvone Suppresses Cellulase-Induced Capsidiol Production in Nicotiana tabacum by Interfering with Protein Isoprenylation. Plant Physiol. 2014, 164, 935-950.