Thermo Scientific Chemicals

Allylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L07162.AC
25 mL, Each
Thermo Scientific Chemicals

Allylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L07162.AC
25 mL, Each
Quantity
Catalog number: L07162.AC
also known as L07162-AC
Price (USD)
Price: 30.40
Your price:
Quantity
-

Chemical Identifiers

CAS
107-11-9
Refractive Index
1.4190-1.4230 @ 20°C (non-U.S. specification)
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms

Description

Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text