Thermo Scientific Chemicals

Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid, Thermo Scientific Chemicals

Catalog number: L07349.30
250 g, Each
Thermo Scientific Chemicals

Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid, Thermo Scientific Chemicals

Catalog number: L07349.30
250 g, Each
Quantity
Catalog number: L07349.30
also known as L07349-30
Price (USD)
Price: 47.90
Online price: 40.65
Your price:
Quantity
-

Chemical Identifiers

CAS
115-27-5
IUPAC Name
1,7,8,9,10,10-hexachloro-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Formula
C9H2Cl6O3
InChI Key
FLBJFXNAEMSXGL-UHFFFAOYNA-N
SMILES
ClC1=C(Cl)C2(Cl)C3C(C(=O)OC3=O)C1(Cl)C2(Cl)Cl
Appearance (Color)
White
Form
Powder and/or Lumps
Assay (Titration ex Anhydride)
>94.0 to <106.0%

Description

Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text