1,2,4-Benzenetricarboxylic acid, 98%
1,2,4-Benzenetricarboxylic acid, 98%
1,2,4-Benzenetricarboxylic acid, 98%
Thermo Scientific Chemicals

1,2,4-Benzenetricarboxylic acid, 98%

CAS: 528-44-9 | C9H6O6 | 210.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07359.14
also known as L07359-14
Price (USD)
50.65
Online Exclusive
56.00
Save 5.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.00
Save 5.35 (10%)
Each
Chemical Identifiers
CAS528-44-9
IUPAC Namebenzene-1,2,4-tricarboxylic acid
Molecular FormulaC9H6O6
InChI KeyARCGXLSVLAOJQL-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder or flakes or lumps
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only