Thermo Scientific Chemicals

Ethidium bromide, 95%, Thermo Scientific Chemicals

Catalog number: L07482.06
5 g, Each
Thermo Scientific Chemicals

Ethidium bromide, 95%, Thermo Scientific Chemicals

Catalog number: L07482.06
5 g, Each
Quantity
Catalog number: L07482.06
also known as L07482-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1239-45-8
IUPAC Name
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular Formula
C21H20BrN3
InChI Key
ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Assay from Supplier's CofA
≥94.0% (U.S. specification)
Form
Crystals or powder or crystalline powder
Appearance (Color)
Red to dark red to purple to dark purple
Assay (Argentometric Titration)
≥94.0 to ≤106.0% (non-U.S. specification)
Loss on Drying
≤5.0% (non-U.S. specification)

Description

For DNA isolation and visualization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text