Methyl trans-2-hexenoate, 97%
Methyl trans-2-hexenoate, 97%
Methyl trans-2-hexenoate, 97%
Methyl trans-2-hexenoate, 97%
Thermo Scientific Chemicals

Methyl trans-2-hexenoate, 97%

CAS: 13894-63-8 | C7H12O2 | 128.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08030.09
also known as L08030-09
Price (TWD)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS13894-63-8
IUPAC Namemethyl (2E)-hex-2-enoate
Molecular FormulaC7H12O2
InChI KeyGFUGBRNILVVWIE-AATRIKPKSA-N
SMILESCCCC=CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4325-1.4365 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only